Tisk…

Allyl-2,2,3,3-tetrafluoropropyl ether 97%

Dodavatel: Apollo Scientific
Nebezpečí

PC9881-100G PC9881-25G PC9881-5G
APOSPC9881-100GEA 20500 CZK
APOSPC9881-100G APOSPC9881-25G APOSPC9881-5G
Allyl-2,2,3,3-tetrafluoropropyl ether 97%
Allyl-2,2,3,3-tetrafluoropropyl ether
Vzorec: C₆H₈F₄O
MW: 172,12 g/mol
Bod varu: 110 °C/740mmHg
Hustota: 1,18 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD02093319
CAS číslo: 681-68-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR