2,2-Difluoro-2-phenylacetonitrile 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC9863-100MG
PC9863-1G
APOSPC9863-100MGEA
622
CZK
APOSPC9863-100MG
APOSPC9863-1G
2,2-Difluoro-2-phenylacetonitrile 95%
2,2-Difluoro-2-phenylacetonitrile
Vzorec:
C₈H₅F₂N Bod varu: 94…95 °C/98mmHg Bod vzplanutí: 54 °C Storage Temperature: Ambient |
MDL Number:
MFCD07784286 CAS číslo: 2002-72-4 UN: 3275 ADR: 6.1+3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...