Tisk…

4-Chloro-3-(trifluoromethyl)pyridine hydrochloride 97%

Dodavatel: Apollo Scientific
Varování

PC9858-1G PC9858-250MG
APOSPC9858-1GEA 4100 CZK
APOSPC9858-1G APOSPC9858-250MG
4-Chloro-3-(trifluoromethyl)pyridine hydrochloride 97%
4-Chloro-3-(trifluoromethyl)pyridine hydrochloride
Vzorec: C₆H₄Cl₂F₃N
MW: 218,01 g/mol
Bod tání: 124…126 °C
Storage Temperature: Ambient
MDL Number: MFCD04972770
CAS číslo: 732306-24-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR