Tritylium tetrafluoroborate
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC9789-25G
PC9789-5G
APOSPC9789-25GEA
5390
CZK
APOSPC9789-25G
APOSPC9789-5G
Tritylium tetrafluoroborate
Tritylium tetrafluoroborate
Vzorec:
C₁₉H₁₅BF₄ Bod tání: 205…215 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00013120 CAS číslo: 341-02-6 EINECS: 206-433-3 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...