3-Fluorosalicylic acid 99%
Dodavatel: Apollo Scientific
|
Varování
|
PC9781-1G
PC9781-25G
PC9781-5G
APOSPC9781-1GEA
622
CZK
APOSPC9781-1G
APOSPC9781-25G
APOSPC9781-5G
3-Fluorosalicylic acid 99%
3-Fluorosalicylic acid
Vzorec:
C₇H₅FO₃ MW: 156,11 g/mol Bod tání: 146…148 °C Storage Temperature: Ambient |
MDL Number:
MFCD00153167 CAS číslo: 341-27-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...