1-Bromo-2-chloro-1,1,2-trifluoroethane 99%
Dodavatel: Apollo Scientific
|
Varování
|
PC9780-100G
PC9780-25G
APOSPC9780-100GEA
10350
CZK
APOSPC9780-100G
APOSPC9780-25G
1-Bromo-2-chloro-1,1,2-trifluoroethane 99%
1-Bromo-2-chloro-1,1,2-trifluoroethane
Vzorec:
BrF₂CCHClF MW: 197,38 g/mol Bod varu: 52…53 °C Hustota: 1,8636 g/cm³ (25 °C) Storage Temperature: Ambient |
CAS číslo:
354-06-3 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...