Tisk…

2-Pyrazin-2-yl-1-[4-(trifluoromethyl)phenyl]ethanone 95+%

Dodavatel: Apollo Scientific

PC9740-1G PC9740-5G
APOSPC9740-1GEA 4630 CZK
APOSPC9740-1G APOSPC9740-5G
2-Pyrazin-2-yl-1-[4-(trifluoromethyl)phenyl]ethanone 95+%
2-Pyrazin-2-yl-1-[4-(trifluoromethyl)phenyl]ethanone
Vzorec: C₁₃H₉F₃N₂O
MW: 266,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04037908
CAS číslo: 88283-34-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR