Tisk…

3-Fluoro-4-(trifluoromethyl)benzamide 98%

Dodavatel: Apollo Scientific
Varování

PC9652-1G PC9652-5G
APOSPC9652-1GEA 622 CZK
APOSPC9652-1G APOSPC9652-5G
3-Fluoro-4-(trifluoromethyl)benzamide 98%
3-Fluoro-4-(trifluoromethyl)benzamide
Vzorec: C₈H₅F₄NO
Storage Temperature: Ambient
CAS číslo: 247170-27-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR