2-(2-Pyridinyl)-6-(trifluoromethyl)-4-pyrimidinethiol
Dodavatel: Apollo Scientific
PC9633-1G
PC9633-500MG
APOSPC9633-1GEA
20500
CZK
APOSPC9633-1G
APOSPC9633-500MG
2-(2-Pyridinyl)-6-(trifluoromethyl)-4-pyrimidinethiol
2-(2-Pyridinyl)-6-(trifluoromethyl)-4-pyrimidinethiol
Vzorec:
C₁₀H₆F₃N₃S Bod tání: 145…148 °C Storage Temperature: Ambient |
MDL Number:
MFCD00139802 CAS číslo: 338418-16-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...