3-Chloro-2-fluorophenylboronic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(3-Chloro-2-fluorophenyl)metaboric acid, Dihydroxy(3-chloro-2-fluorophenyl)borane, 3-Chloro-2-fluorophenylboronic Acid, 3-Chloro-2-fluorobenzeneboronic acid
PC9594-1G
PC9594-5G
PC9594-25G
APOSPC9594-1GEA
622
CZK
APOSPC9594-1G
APOSPC9594-5G
APOSPC9594-25G
3-Chloro-2-fluorophenylboronic acid
3-Chloro-2-fluorophenylboronic acid
Vzorec:
C₆H₅BClFO₂ Bod tání: 223 °C Storage Temperature: Ambient |
MDL Number:
MFCD05664224 CAS číslo: 352535-82-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...