4-(Trifluoromethyl)phenylacetylene 99%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
4-Ethynylbenzotrifluoride, 4-Ethynyl-alpha,alpha,alpha-trifluorotoluene, 1-ethynyl-4-(trifluoromethyl)benzene
PC9586-25G
PC9586-1G
PC9586-5G
APOSPC9586-25GEA
7080
CZK
APOSPC9586-25G
APOSPC9586-1G
APOSPC9586-5G
4-(Trifluoromethyl)phenylacetylene 99%
4-(Trifluoromethyl)phenylacetylene
Vzorec:
C₉H₅F₃ MW: 170.134 g/mol Bod varu: 78…80 °C/2mmHg Hustota: 1.043 g/cm³ (25 °C) Bod vzplanutí: 20.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD01861903 CAS číslo: 705-31-7 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...