2-Chloro-5-fluoro-3-methylpyridine 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Chloro-5-fluoro-3-picoline
PC9553-1G
PC9553-500MG
APOSPC9553-1GEA
827
CZK
APOSPC9553-1G
APOSPC9553-500MG
2-Chloro-5-fluoro-3-methylpyridine 98%
2-Chloro-5-fluoro-3-methylpyridine
Vzorec:
C₆H₅ClFN MW: 145.56 Bod tání: 43…46 °C Storage Temperature: Ambient |
MDL Number:
MFCD04972725 CAS číslo: 38186-84-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...