2-Methoxy-2-phenyl-3,3,3-trifluoropropionitrile
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
alpha-Methoxy-alpha-(trifluoromethyl)phenylacetonitrile
PC9549-1G
PC9549-5G
APOSPC9549-1GEA
417
CZK
APOSPC9549-1G
APOSPC9549-5G
2-Methoxy-2-phenyl-3,3,3-trifluoropropionitrile
2-Methoxy-2-phenyl-3,3,3-trifluoropropionitrile
Vzorec:
C₁₀H₈F₃NO MW: 215,18 g/mol Bod varu: 76…82 °C/12mmHg Hustota: 1,152 g/cm³ (25 °C) Bod vzplanutí: 89 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001844 CAS číslo: 80866-87-1 UN: 3276 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...