2-Chloro-3-fluoropyridine 99%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC9542-25G
PC9542-5G
PC9542-100G
APOSPC9542-25GEA
705
CZK
APOSPC9542-25G
APOSPC9542-5G
APOSPC9542-100G
2-Chloro-3-fluoropyridine 99%
2-Chloro-3-fluoropyridine
Vzorec:
C₅H₃ClFN MW: 131,54 g/mol Bod varu: 80 °C/80mmHg Hustota: 1,323 g/cm³ (25 °C) Bod vzplanutí: 63 °C Storage Temperature: Ambient |
MDL Number:
MFCD03095302 CAS číslo: 17282-04-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...