Tisk…

1-{[3-(Trifluoromethyl)phenyl]sulphonyl}-2-pyrrolidinecarboxylic acid 97%

Dodavatel: Apollo Scientific

PC9504-1G PC9504-500MG PC9504-5G
APOSPC9504-1GEA 4900 CZK
APOSPC9504-1G APOSPC9504-500MG APOSPC9504-5G
1-{[3-(Trifluoromethyl)phenyl]sulphonyl}-2-pyrrolidinecarboxylic acid 97%
1-{[3-(Trifluoromethyl)phenyl]sulphonyl}-2-pyrrolidinecarboxylic acid
Vzorec: C₁₂H₁₂F₃NO₄S
MW: 323,29 g/mol
Bod tání: 107…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00172809
CAS číslo: 251096-97-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR