3-{[3-(Trifluoromethyl)benzyl]sulphanyl}-1H-1,2,4-triazol-5-amine 97%
Dodavatel: Apollo Scientific
Synonyms:
5-Amino-3-[3-(trifluoromethyl)benzylthio]-1H-1,2,4-triazole
PC9492-1G
PC9492-500MG
PC9492-5G
APOSPC9492-1GEA
7840
CZK
APOSPC9492-1G
APOSPC9492-500MG
APOSPC9492-5G
3-{[3-(Trifluoromethyl)benzyl]sulphanyl}-1H-1,2,4-triazol-5-amine 97%
3-{[3-(Trifluoromethyl)benzyl]sulphanyl}-1H-1,2,4-triazol-5-amine
Vzorec:
C₁₀H₉F₃N₄S MW: 274,27 g/mol Bod tání: 136…138 °C Storage Temperature: Ambient |
MDL Number:
MFCD00141527 CAS číslo: 303150-56-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...