2-Oxo-1-[4-(trifluoromethyl)benzyl]-1,2-dihydro-3-pyridine carboxylic acid 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1-[4-(Trifluoromethyl)benzyl]pyridin-2-one-3-carboxylic acid
PC9473-1G
PC9473-5G
APOSPC9473-1GEA
1660
CZK
APOSPC9473-1G
APOSPC9473-5G
2-Oxo-1-[4-(trifluoromethyl)benzyl]-1,2-dihydro-3-pyridine carboxylic acid 97%
2-Oxo-1-[4-(trifluoromethyl)benzyl]-1,2-dihydro-3-pyridine carboxylic acid
Vzorec:
C₁₄H₁₀F₃NO₃ MW: 297.24 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00243696 CAS číslo: 66158-46-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...