Tisk…

3-Nitro-4-[3-(trifluoromethyl)phenoxy]benzenecarbaldehyde 97%

Dodavatel: Apollo Scientific

PC9467-1G PC9467-500MG PC9467-5G
APOSPC9467-1GEA 7840 CZK
APOSPC9467-1G APOSPC9467-500MG APOSPC9467-5G
3-Nitro-4-[3-(trifluoromethyl)phenoxy]benzenecarbaldehyde 97%
3-Nitro-4-[3-(trifluoromethyl)phenoxy]benzenecarbaldehyde
Vzorec: C₁₄H₈F₃NO₄
Bod tání: 107…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00138749
CAS číslo: 320416-66-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR