Tisk…

{1-Methyl-3-(trifluoromethyl)-5-[3-(trifluoromethyl)phenoxy]-1H-pyrazol-4-yl}methanol 97%

Dodavatel: Apollo Scientific

PC9464-1G PC9464-500MG PC9464-5G
APOSPC9464-1GEA 15180 CZK
APOSPC9464-1G APOSPC9464-500MG APOSPC9464-5G
{1-Methyl-3-(trifluoromethyl)-5-[3-(trifluoromethyl)phenoxy]-1H-pyrazol-4-yl}methanol 97%
{1-Methyl-3-(trifluoromethyl)-5-[3-(trifluoromethyl)phenoxy]-1H-pyrazol-4-yl}methanol
Vzorec: C₁₃H₁₀F₆N₂O₂
Bod tání: 81…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00172663
CAS číslo: 318469-40-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR