3-Methyl-2-({[3-(trifluoromethyl)phenyl]sulphonyl}amino)butanoic acid 97%
Dodavatel: Apollo Scientific
Synonyms:
3-Methyl-2-[3-(trifluoromethyl)benzenesulphonylamino]butyric acid
PC9455-1G
PC9455-500MG
PC9455-5G
APOSPC9455-1GEA
7840
CZK
APOSPC9455-1G
APOSPC9455-500MG
APOSPC9455-5G
3-Methyl-2-({[3-(trifluoromethyl)phenyl]sulphonyl}amino)butanoic acid 97%
3-Methyl-2-({[3-(trifluoromethyl)phenyl]sulphonyl}amino)butanoic acid
Vzorec:
C₁₂H₁₄F₃NO₄S MW: 325,31 g/mol Bod tání: 127…129 °C Storage Temperature: Ambient |
MDL Number:
MFCD00173010 CAS číslo: 250714-85-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...