2-Methyl-N-[3-(trifluoromethyl)phenyl]-2-oxiranecarboxamide 97%
Dodavatel: Apollo Scientific
PC9448-1G
PC9448-500MG
PC9448-5G
APOSPC9448-1GEA
7840
CZK
APOSPC9448-1G
APOSPC9448-500MG
APOSPC9448-5G
2-Methyl-N-[3-(trifluoromethyl)phenyl]-2-oxiranecarboxamide 97%
2-Methyl-N-[3-(trifluoromethyl)phenyl]-2-oxiranecarboxamide
Vzorec:
C₁₁H₁₀F₃NO₂ MW: 245,2 g/mol Bod tání: 62…66 °C Storage Temperature: Ambient |
MDL Number:
MFCD00232083 CAS číslo: 338424-38-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...