3-Amino-4-fluorobenzoic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-Amino-4-fluorobenzoic Acid
PC9442-5G
PC9442-100G
PC9442-25G
APOSPC9442-5GEA
622
CZK
APOSPC9442-5G
APOSPC9442-100G
APOSPC9442-25G
3-Amino-4-fluorobenzoic acid
3-Amino-4-fluorobenzoic acid
Vzorec:
C₇H₆FNO₂ Bod tání: 182…183 °C Storage Temperature: Ambient |
MDL Number:
MFCD00130055 CAS číslo: 2365-85-7 EINECS: 219-122-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...