2',6'-Dibromo-4'-(trifluoromethyl)acetanilide
Dodavatel: Apollo Scientific
PC9425-1G
PC9425-500MG
APOSPC9425-1GEA
6550
CZK
APOSPC9425-1G
APOSPC9425-500MG
2',6'-Dibromo-4'-(trifluoromethyl)acetanilide
2',6'-Dibromo-4'-(trifluoromethyl)acetanilide
Vzorec:
C₉H₆Br₂F₃NO Storage Temperature: Ambient |
MDL Number:
MFCD03094553 CAS číslo: 544716-06-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...