Tisk…

Methyl-4-[2-nitro-4-(trifluoromethyl)anilino]butanoate 97%

Dodavatel: Apollo Scientific

PC9424-10G PC9424-1G PC9424-5G
APOSPC9424-10GEA 15540 CZK
APOSPC9424-10G APOSPC9424-1G APOSPC9424-5G
Methyl-4-[2-nitro-4-(trifluoromethyl)anilino]butanoate 97%
Methyl-4-[2-nitro-4-(trifluoromethyl)anilino]butanoate
Vzorec: C₁₂H₁₃F₃N₂O₄
Bod tání: 81…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00793100
CAS číslo: 339101-25-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR