Tisk…

Methyl-3-amino-1-cyano-4-fluoro-1-phenyl-1H-indene-2-carboxylate 97%

Dodavatel: Apollo Scientific

PC9390-1G PC9390-500MG PC9390-5G
APOSPC9390-1GEA 11540 CZK
APOSPC9390-1G APOSPC9390-500MG APOSPC9390-5G
Methyl-3-amino-1-cyano-4-fluoro-1-phenyl-1H-indene-2-carboxylate 97%
Methyl-3-amino-1-cyano-4-fluoro-1-phenyl-1H-indene-2-carboxylate
Vzorec: C₁₈H₁₃FN₂O₂
MW: 308,31 g/mol
Bod tání: 154…156 °C
Storage Temperature: Ambient
MDL Number: MFCD00232259
CAS číslo: 339010-38-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR