Tisk…

2-Hydrazino-2-oxo-N-[3-(trifluoromethyl)phenyl]acetamide

Dodavatel: Apollo Scientific

Synonyms: Oxalic monohydrazide mono(3-trifluoromethyl)anilide

PC9383-1G PC9383-500MG PC9383-5G
APOSPC9383-1GEA 7840 CZK
APOSPC9383-1G APOSPC9383-500MG APOSPC9383-5G
2-Hydrazino-2-oxo-N-[3-(trifluoromethyl)phenyl]acetamide
2-Hydrazino-2-oxo-N-[3-(trifluoromethyl)phenyl]acetamide
Vzorec: C₉H₈F₃N₃O₂
MW: 247,18 g/mol
Bod tání: 199…201 °C
Storage Temperature: Ambient
MDL Number: MFCD00122710
CAS číslo: 324016-20-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR