Tisk…

2-Hydrazino-2-oxo-N-[4-(trifluoromethoxy)phenyl]acetamide

Dodavatel: Apollo Scientific

Synonyms: Oxalic monohydrazide mono(4-trifluoromethoxy)anilide

PC9382-1G PC9382-500MG PC9382-5G
APOSPC9382-1GEA 7840 CZK
APOSPC9382-1G APOSPC9382-500MG APOSPC9382-5G
2-Hydrazino-2-oxo-N-[4-(trifluoromethoxy)phenyl]acetamide
2-Hydrazino-2-oxo-N-[4-(trifluoromethoxy)phenyl]acetamide
Vzorec: C₉H₈F₃N₃O₃
MW: 263,18 g/mol
Bod tání: 215…217 °C
Storage Temperature: Ambient
MDL Number: MFCD00173093
CAS číslo: 338405-24-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR