α-Bromo-4-chloro-3-fluorotoluene 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC9381-25G
PC9381-10G
PC9381-100G
PC9381-5G
APOSPC9381-25GEA
3110
CZK
APOSPC9381-25G
APOSPC9381-10G
APOSPC9381-100G
APOSPC9381-5G
α-Bromo-4-chloro-3-fluorotoluene 97%
α-Bromo-4-chloro-3-fluorotoluene
Vzorec:
C₇H₅BrClF Bod tání: 30…33 °C Storage Temperature: Ambient |
MDL Number:
MFCD04115859 CAS číslo: 206362-80-3 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...