[5-[(4-Fluorophenyl)sulphanyl]-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol 97%
Dodavatel: Apollo Scientific
PC9370-1G
PC9370-500MG
PC9370-5G
APOSPC9370-1GEA
15180
CZK
APOSPC9370-1G
APOSPC9370-500MG
APOSPC9370-5G
[5-[(4-Fluorophenyl)sulphanyl]-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol 97%
[5-[(4-Fluorophenyl)sulphanyl]-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol
Vzorec:
C₁₂H₁₀F₄N₂OS Bod tání: 66…67 °C Storage Temperature: Ambient |
MDL Number:
MFCD00172648 CAS číslo: 318239-65-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...