Tisk…

5-[(4-Fluorophenyl)sulphanyl]-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde 97%

Dodavatel: Apollo Scientific

PC9368-1G PC9368-500MG
APOSPC9368-1GEA 14520 CZK
APOSPC9368-1G APOSPC9368-500MG
5-[(4-Fluorophenyl)sulphanyl]-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde 97%
5-[(4-Fluorophenyl)sulphanyl]-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde
Vzorec: C₁₂H₈F₄N₂OS
MW: 304,27 g/mol
Bod tání: 61…62 °C
Storage Temperature: Ambient
MDL Number: MFCD01935982
CAS číslo: 321848-46-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR