N-(4-Fluorophenyl)-2-hydrazino-2-oxoacetamide
Dodavatel: Apollo Scientific
PC9356-1G
PC9356-500MG
PC9356-5G
APOSPC9356-1GEA
7840
CZK
APOSPC9356-1G
APOSPC9356-500MG
APOSPC9356-5G
N-(4-Fluorophenyl)-2-hydrazino-2-oxoacetamide
N-(4-Fluorophenyl)-2-hydrazino-2-oxoacetamide
Vzorec:
C₈H₈FN₃O₂ Bod tání: 223…226 °C Storage Temperature: Ambient |
MDL Number:
MFCD00119047 CAS číslo: 338395-87-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...