2-(4-Fluorophenoxy)-1-methyl-1H-indole-3-carbaldehyde 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC9349-1G
PC9349-500MG
APOSPC9349-1GEA
16070
CZK
APOSPC9349-1G
APOSPC9349-500MG
2-(4-Fluorophenoxy)-1-methyl-1H-indole-3-carbaldehyde 97%
2-(4-Fluorophenoxy)-1-methyl-1H-indole-3-carbaldehyde
Vzorec:
C₁₆H₁₂FNO₂ MW: 269,28 g/mol Bod tání: 139…141 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01315828 CAS číslo: 338416-70-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...