Tisk…

2,2-Difluoro-5-cyano-1,3-benzodioxole 96%

Dodavatel: Apollo Scientific

Synonyms: 2,2-Difluoro-1,3-benzodioxole-5-carbonitrile

PC9343-1G PC9343-5G
APOSPC9343-1GEA 3740 CZK
APOSPC9343-1G APOSPC9343-5G
2,2-Difluoro-5-cyano-1,3-benzodioxole 96%
2,2-Difluoro-5-cyano-1,3-benzodioxole
Vzorec: C₈H₃F₂NO₂
MW: 183,12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00792418
CAS číslo: 135132-34-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR