4-[(4-Fluorobenzyl)oxy]-3-methoxybenzenecarbaldehyde 97%
Dodavatel: Apollo Scientific
Synonyms:
4-((4-Fluorobenzyl)oxy)-3-methoxybenzaldehyde
PC9333-10G
PC9333-1G
PC9333-5G
APOSPC9333-10GEA
21560
CZK
APOSPC9333-10G
APOSPC9333-1G
APOSPC9333-5G
4-[(4-Fluorobenzyl)oxy]-3-methoxybenzenecarbaldehyde 97%
4-[(4-Fluorobenzyl)oxy]-3-methoxybenzenecarbaldehyde
Vzorec:
C₁₅H₁₃FO₃ MW: 260.27 g/mol Bod tání: 80…82 °C Storage Temperature: Ambient |
MDL Number:
MFCD00202628 CAS číslo: 321432-05-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...