4-Fluorodiphenylmethane 95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1-Benzyl-4-fluorobenzene
PC9325-25G
PC9325-5G
APOSPC9325-25GEA
16300
CZK
APOSPC9325-25G
APOSPC9325-5G
4-Fluorodiphenylmethane 95%
4-Fluorodiphenylmethane
Vzorec:
C₁₃H₁₁F Bod varu: 132 °C Hustota: 1,09 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00464329 CAS číslo: 587-79-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...