Tisk…

Ytterbium(III) trifluoromethanesulphonate 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: Ytterbium (III) triflate

PC9300-25G PC9300-5G PC9300-100G
APOSPC9300-25GEA 5790 CZK
APOSPC9300-25G APOSPC9300-5G APOSPC9300-100G
Ytterbium(III) trifluoromethanesulphonate 97%
Ytterbium(III) trifluoromethanesulphonate
Vzorec: C₃F₉O₉S₃Yb
MW: 620.25
Storage Temperature: Ambient
MDL Number: MFCD06200261
CAS číslo: 54761-04-5
UN: 3261
ADR: 8 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR