2,6-Difluorophenylboronic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
PC9293-25G
PC9293-5G
APOSPC9293-25GEA
1870
CZK
APOSPC9293-25G
APOSPC9293-5G
2,6-Difluorophenylboronic acid 98%
2,6-Difluorophenylboronic acid
Vzorec:
C₆H₅BF₂O₂ Bod tání: 147…149 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00792436 CAS číslo: 162101-25-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...