Tisk…

2-Chloro-3-(trifluoromethyl)benzyl bromide 97%

Dodavatel: Apollo Scientific
Nebezpečí

PC9289-5G PC9289-1G PC9289-25G
APOSPC9289-5GEA 3230 CZK
APOSPC9289-5G APOSPC9289-1G APOSPC9289-25G
2-Chloro-3-(trifluoromethyl)benzyl bromide 97%
2-Chloro-3-(trifluoromethyl)benzyl bromide
Vzorec: C₈H₅BrClF₃
MW: 273.48
Storage Temperature: Ambient
MDL Number: MFCD01631539
CAS číslo: 261763-22-8
UN: 3265
ADR: 8 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR