Tisk…

2-Fluoro-6-methylbenzaldehyde 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Fluoro-6-tolualdehyde, 6-Fluoro-o-tolualdehyde, 6-Fluoro-o-tolylaldehyde

PC9285-1G PC9285-25G PC9285-5G
APOSPC9285-1GEA 1330 CZK
APOSPC9285-1G APOSPC9285-25G APOSPC9285-5G
2-Fluoro-6-methylbenzaldehyde 98%
2-Fluoro-6-methylbenzaldehyde
Vzorec: C₈H₇FO
MW: 138.14
Storage Temperature: Ambient
MDL Number: MFCD09258962
CAS číslo: 117752-04-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR