Tisk…

3-Bromo-5-fluorophenylacetonitrile 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-(3-bromo-5-fluorophenyl)ethanenitrile, Benzeneacetonitrile, 3-bromo-5-fluoro-, 2-(3-bromo-5-fluorophenyl)acetonitrile

PC9277-1G PC9277-5G
APOSPC9277-1GEA 2150 CZK
APOSPC9277-1G APOSPC9277-5G
3-Bromo-5-fluorophenylacetonitrile 98%
3-Bromo-5-fluorophenylacetonitrile
Vzorec: C₈H₅BrFN
MW: 214,037 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09258960
CAS číslo: 305800-58-2
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR