Tisk…

3-Nitro-5-(trifluoromethyl)benzonitrile 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 3-Cyano-5-nitrobenzotrifluoride

PC9255-5G PC9255-1G PC9255-25G
APOSPC9255-5GEA 827 CZK
APOSPC9255-5G APOSPC9255-1G APOSPC9255-25G
3-Nitro-5-(trifluoromethyl)benzonitrile 97%
3-Nitro-5-(trifluoromethyl)benzonitrile
Vzorec: C₈H₃F₃N₂O₂
MW: 216,1200 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03412207
CAS číslo: 20566-80-7
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR