Tisk…

(2-Fluoro-4-methoxyphenyl)acetic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Fluoro-4-methoxyphenylacetic acid

PC9191-1G PC9191-5G
APOSPC9191-1GEA 705 CZK
APOSPC9191-1G APOSPC9191-5G
(2-Fluoro-4-methoxyphenyl)acetic acid
(2-Fluoro-4-methoxyphenyl)acetic acid
Vzorec: C₉H₉FO₃
MW: 184,17 g/mol
Bod tání: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD06800744
CAS číslo: 883531-28-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR