Tisk…

4-Fluoro-3-nitrophenylboronic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Fluoro-3-nitrobenzeneboronic acid

PC9161-1G PC9161-5G
APOSPC9161-1GEA 2900 CZK
APOSPC9161-1G APOSPC9161-5G
4-Fluoro-3-nitrophenylboronic acid
4-Fluoro-3-nitrophenylboronic acid
Vzorec: C₆H₅BFNO₄
Storage Temperature: Refrigerator
MDL Number: MFCD08277236
CAS číslo: 352530-22-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR