Tisk…

2-Bromo-4-(trifluoromethyl)phenylboronic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Bromo-4-(trifluoromethyl)benzeneboronic acid, (2-Bromo-4-(trifluoromethyl)phenyl)boronic acid

PC9160-1G PC9160-5G
APOSPC9160-1GEA 1660 CZK
APOSPC9160-1G APOSPC9160-5G
2-Bromo-4-(trifluoromethyl)phenylboronic acid
2-Bromo-4-(trifluoromethyl)phenylboronic acid
Vzorec: C₇H₅BBrF₃O₂
MW: 268.82
Storage Temperature: Freezer
MDL Number: MFCD08532500
CAS číslo: 959997-88-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR