2-Bromo-4-(trifluoromethyl)quinoline
Dodavatel: Apollo Scientific
PC9154-1G
PC9154-250MG
PC9154-500MG
PC9154-2.5G
APOSPC9154-1GEA
2860
CZK
APOSPC9154-1G
APOSPC9154-250MG
APOSPC9154-500MG
APOSPC9154-2.5G
2-Bromo-4-(trifluoromethyl)quinoline
2-Bromo-4-(trifluoromethyl)quinoline
Vzorec:
C₁₀H₅BrF₃N Storage Temperature: Ambient |
MDL Number:
MFCD08458129 CAS číslo: 590372-17-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...