4-(Trifluoromethyl)quinoline-2-carboxylic acid
Dodavatel: Apollo Scientific
PC9151-1G
PC9151-50MG
PC9151-250MG
PC9151-500MG
APOSPC9151-1GEA
10820
CZK
APOSPC9151-1G
APOSPC9151-50MG
APOSPC9151-250MG
APOSPC9151-500MG
4-(Trifluoromethyl)quinoline-2-carboxylic acid
4-(Trifluoromethyl)quinoline-2-carboxylic acid
Vzorec:
C₁₁H₆F₃NO₂ MW: 241,17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06796588 CAS číslo: 588702-67-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...