3-(Trifluoromethyl)quinoline-4-carboxylic acid
Dodavatel: Apollo Scientific
|
Varování
|
PC9148-1G
PC9148-50MG
PC9148-250MG
PC9148-500MG
PC9148-2.5G
PC9148-5G
APOSPC9148-1GEA
12300
CZK
APOSPC9148-1G
APOSPC9148-50MG
APOSPC9148-250MG
APOSPC9148-500MG
APOSPC9148-2.5G
APOSPC9148-5G
3-(Trifluoromethyl)quinoline-4-carboxylic acid
3-(Trifluoromethyl)quinoline-4-carboxylic acid
Vzorec:
C₁₁H₆F₃NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD08458858 CAS číslo: 588702-65-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...