Tisk…

1,1,2-Trifluoro-2-chloro-3-methyl-3-vinylcyclobutane

Dodavatel: Apollo Scientific
Varování

PC9135-25G PC9135-5G
APOSPC9135-25GEA 3280 CZK
APOSPC9135-25G APOSPC9135-5G
1,1,2-Trifluoro-2-chloro-3-methyl-3-vinylcyclobutane
1,1,2-Trifluoro-2-chloro-3-methyl-3-vinylcyclobutane
Vzorec: C₇H₈ClF₃
MW: 184,59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00039438
CAS číslo: 4265-28-5
UN: 1993
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR