Tisk…

3-(Trifluoromethyl)picolinic acid 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 3-(Trifluoromethyl)pyridine-2-carboxylic acid, 3-(Trifluoromethy)pyridine-2-carboxylic acid

PC9130-1G PC9130-5G PC9130-10G
APOSPC9130-1GEA 622 CZK
APOSPC9130-1G APOSPC9130-5G APOSPC9130-10G
3-(Trifluoromethyl)picolinic acid 97%
3-(Trifluoromethyl)picolinic acid
Vzorec: C₇H₄F₃NO₂
Bod tání: 125…130 °C
Storage Temperature: Ambient
MDL Number: MFCD07375386
CAS číslo: 87407-12-3
UN: 2811
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR