4-(Bromodifluoromethyl)-1-(difluoromethyl)benzene 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-[Bromo(difluoro)methyl]benzal fluoride
PC9124-1G
PC9124-5G
PC9124-250MG
APOSPC9124-1GEA
11540
CZK
APOSPC9124-1G
APOSPC9124-5G
APOSPC9124-250MG
4-(Bromodifluoromethyl)-1-(difluoromethyl)benzene 97%
4-(Bromodifluoromethyl)-1-(difluoromethyl)benzene
Vzorec:
C₈H₅BrF₄ MW: 257,03 g/mol Bod varu: 59 °C/3mmHg Storage Temperature: Ambient |
MDL Number:
MFCD03094540 CAS číslo: 2250-36-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...